Learn More
TARGETMOL CHEMICALS INC ISTRADEFYLLINE 100MG
SDP
Catalog No. 502579335
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 50 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Istradefylline (KW-6002)(Ki of 2.2 nM) is a selective adenosine A2A receptor (A2AR) antagonist which is under development in Phase 3 trails. It has been used in trials studying the treatment and basic science of Drug Abuse Sleep Disorder Hepatic Impairment Parkinson's Disease and Restless Legs Syndrome among others. purity: 98%
Catalog No. 50-257-9335 Supplier TARGETMOL CHEMICALS INC Supplier No. T6552100MG
Also available in 5 mg 10 mg 25 mg 50 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Istradefylline (KW-6002)(Ki of 2.2 nM) is a selective adenosine A2A receptor (A2AR) antagonist which is under development in Phase 3 trails. It has been used in trials studying the treatment and basic science of Drug Abuse Sleep Disorder Hepatic Impairment Parkinson's Disease and Restless Legs Syndrome among others. purity: 98%