Learn More
Medchemexpress LLC Istradefylline-13C,d3 | 2749234-46-4 | 98.3% | 388.44 | 10 MG
SDP
Catalog No. 5000345835
Encompass_Preferred
Istradefylline-13C,d3 is the 13C- and deuterium labeled version of Istradefylline, a highly potent, selective, and orally active adenosine A2A receptor antagonist with a Ki of 2.2 nM. It has been studied in experimental models of Parkinson's disease. This compound is intended for research use only and can be used for quantitative analysis.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-45835 Supplier Medchemexpress LLC Supplier No. HY10888S10MG
Istradefylline-13C,d3 is the 13C- and deuterium labeled version of Istradefylline, a highly potent, selective, and orally active adenosine A2A receptor antagonist with a Ki of 2.2 nM. It has been studied in experimental models of Parkinson's disease. This compound is intended for research use only and can be used for quantitative analysis.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS