Learn More
Medchemexpress LLC Istradefylline-13C,d3 | 2749234-46-4 | 98.3% | 388.44 | 50 MG
SDP
Catalog No. 5000346113
Encompass_Preferred
Istradefylline-13C,d3 is the 13C- and deuterium labeled Istradefylline. It is a very potent, selective and orally active adenosine A2A receptor antagonist with Ki of 2.2 nM in experimental models of Parkinson's disease.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-46113 Supplier Medchemexpress LLC Supplier No. HY10888S1MG
Istradefylline-13C,d3 is the 13C- and deuterium labeled Istradefylline. It is a very potent, selective and orally active adenosine A2A receptor antagonist with Ki of 2.2 nM in experimental models of Parkinson's disease.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS