Learn More
Medchemexpress LLC Istradefylline-13C,d3 | 2749234-46-4 | 98.3% | 388.44 g/mol | C1913CH21D3N4O4 | 5 MG
SDP
Catalog No. 5000263678
Encompass_Preferred
Istradefylline-13C,d3 is a 13C- and deuterium-labeled analog of istradefylline intended for use as a stable isotope internal standard in quantitative mass spectrometry and analytical workflows. It is supplied as a light-yellow solid with high reported chemical purity and packaged in small, laboratory-scale amounts for precision use.
  • Stable isotope-labeled internal standard for LC-MS/GC-MS quantification.
  • High chemical purity suitable for analytical applications.
  • Labeled with 13C and deuterium for clear mass separation from analyte.
  • Supplied as a solid for easy handling and storage.
  • Available in small, precise quantities for method development and validation.
Catalog No. 50-002-63678 Supplier Medchemexpress LLC Supplier No. HY10888S5MG
Istradefylline-13C,d3 is a 13C- and deuterium-labeled analog of istradefylline intended for use as a stable isotope internal standard in quantitative mass spectrometry and analytical workflows. It is supplied as a light-yellow solid with high reported chemical purity and packaged in small, laboratory-scale amounts for precision use.
  • Stable isotope-labeled internal standard for LC-MS/GC-MS quantification.
  • High chemical purity suitable for analytical applications.
  • Labeled with 13C and deuterium for clear mass separation from analyte.
  • Supplied as a solid for easy handling and storage.
  • Available in small, precise quantities for method development and validation.