Learn More
Selleck Chemical LLC Istradefylline 5mg 155270-99-8 KW-6002
SDP
Catalog No. 501362967
Encompass_Preferred
Istradefylline (KW-6002) is a selective adenosine A2A receptor (A2AR) antagonist with Ki of 2.2 nM. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-2967 Supplier Selleck Chemical LLC Supplier No. S27905MG
Istradefylline (KW-6002) is a selective adenosine A2A receptor (A2AR) antagonist with Ki of 2.2 nM. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.