Learn More
Medchemexpress LLC Istradefylline (Standard) | 155270-99-8 | 99.8% | 384.43 | 1 MG
SDP
Catalog No. 5000345363
Encompass_Preferred
Istradefylline (Standard) is the analytical standard of Istradefylline, intended for research and analytical applications. It is a potent, selective, and orally active adenosine A2A receptor antagonist with a Ki of 2.2 nM in experimental models of Parkinson's disease.
  • Analytical standard for Istradefylline
  • Intended for research and analytical applications
  • Potent, selective, and orally active adenosine A2A receptor antagonist
  • Used in experimental models of Parkinson's disease
Catalog No. 50-003-45363 Supplier Medchemexpress LLC Supplier No. HY10888R100MG
Istradefylline (Standard) is the analytical standard of Istradefylline, intended for research and analytical applications. It is a potent, selective, and orally active adenosine A2A receptor antagonist with a Ki of 2.2 nM in experimental models of Parkinson's disease.
  • Analytical standard for Istradefylline
  • Intended for research and analytical applications
  • Potent, selective, and orally active adenosine A2A receptor antagonist
  • Used in experimental models of Parkinson's disease