Learn More
Medchemexpress LLC Istradefylline (Standard) | 155270-99-8 | 99.8% | 384.43 | 1 MG
SDP
Catalog No. 5000345789
Encompass_Preferred
Istradefylline (Standard) is the analytical standard of Istradefylline. This product is intended for research and analytical applications. Istradefylline is a very potent, selective and orally active adenosine A2A receptor antagonist with Ki of 2.2 nM in experimental models of Parkinson's disease. The compound is an analytical standard used as a reference standard in assays. It is commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Analytical standard for research and analytical applications
  • Potent, selective, and orally active adenosine A2A receptor antagonist
  • Used as a reference standard in assays
  • Suitable for qualitative, quantitative, and methodological research experiments
  • Applicable in HPLC, GC, and MS
Catalog No. 50-003-45789 Supplier Medchemexpress LLC Supplier No. HY10888R10MG
Istradefylline (Standard) is the analytical standard of Istradefylline. This product is intended for research and analytical applications. Istradefylline is a very potent, selective and orally active adenosine A2A receptor antagonist with Ki of 2.2 nM in experimental models of Parkinson's disease. The compound is an analytical standard used as a reference standard in assays. It is commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Analytical standard for research and analytical applications
  • Potent, selective, and orally active adenosine A2A receptor antagonist
  • Used as a reference standard in assays
  • Suitable for qualitative, quantitative, and methodological research experiments
  • Applicable in HPLC, GC, and MS