Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Istradefylline (standard) | 155270-99-8 | MFCD00928421 | 99.8% | 384.43 | C20H24N4O4 | 5 MG
SDP

Catalog No. 5000263437
Encompass_Preferred

Istradefylline (Standard) is an analytical standard intended for research and analytical applications. It is a potent, selective adenosine A2A receptor antagonist (Ki ≈ 2.2 nM) used in Parkinson's disease pharmacology, assay development, and method validation. The material is supplied as a solid at high purity for use as a reference standard and for quality-control purposes.

  • High purity suitable for analytical reference.
  • Molecular weight 384.43 and formula C20H24N4O4.
  • Available in milligram package sizes for standards and assays.
  • Solid, off-white to light yellow physical form for convenient handling.
  • Supplied with certificate of analysis and safety documentation.

Catalog No. 50-002-63437 Supplier Medchemexpress LLC Supplier No. HY10888R5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Istradefylline (Standard) is an analytical standard intended for research and analytical applications. It is a potent, selective adenosine A2A receptor antagonist (Ki ≈ 2.2 nM) used in Parkinson's disease pharmacology, assay development, and method validation. The material is supplied as a solid at high purity for use as a reference standard and for quality-control purposes.

  • High purity suitable for analytical reference.
  • Molecular weight 384.43 and formula C20H24N4O4.
  • Available in milligram package sizes for standards and assays.
  • Solid, off-white to light yellow physical form for convenient handling.
  • Supplied with certificate of analysis and safety documentation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.