Learn More
Medchemexpress LLC HY-12323 10mg , ISX-9 CAS:832115-62-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1232310MG
Encompass_Preferred
HY-12323 10mg ISX-9 CAS: 832115-62-5 ISX-9 (Isoxazole 9) is a potent inducer of adult neural stem cell differentiation. ISX-9 activates Ca2+ influx through both voltage-gated Ca2+ channels and NMDA receptors and increases neuroD expression. ISX-9 also induces cardiomyogenic differentiation of Notch-activated epicardium-derived cells (NECs). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9324