Learn More
Aviva Systems Biology Corp ISYNA1 Antibody - N-terminal region (ARP53716_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53716_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ISYNA1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ISYNA1. Alternate names: IPS, INO1, INOS, IPS 1, IPS-1ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rat, Yeast
Catalog No. 50-014-61104