Learn More
Medchemexpress LLC IT1t dihydrochloride | 1092776-63-0 | 99.8% | 479.57 | C21H36Cl2N4S2 | 5 MG
SDP
Catalog No. 501932139
Encompass_Preferred
IT1t dihydrochloride is a research-grade small-molecule antagonist of the chemokine receptor CXCR4. It inhibits CXCL12/CXCR4 interaction with high potency and is commonly used in receptor signaling and antiviral assays. The compound is supplied as the dihydrochloride salt in solid and solution formats for in vitro studies.
  • Potent CXCR4 antagonist with IC50 ≈ 2.1 nM.
  • High purity (≈99.8%) suitable for in vitro studies.
  • Supplied as dihydrochloride salt for improved stability.
  • Available as solid and 10 mM solution in DMSO.
  • Suitable for receptor binding, signaling, and antiviral assays.
Catalog No. 50-193-2139 Supplier Medchemexpress LLC Supplier No. HY101458A5MG
IT1t dihydrochloride is a research-grade small-molecule antagonist of the chemokine receptor CXCR4. It inhibits CXCL12/CXCR4 interaction with high potency and is commonly used in receptor signaling and antiviral assays. The compound is supplied as the dihydrochloride salt in solid and solution formats for in vitro studies.
  • Potent CXCR4 antagonist with IC50 ≈ 2.1 nM.
  • High purity (≈99.8%) suitable for in vitro studies.
  • Supplied as dihydrochloride salt for improved stability.
  • Available as solid and 10 mM solution in DMSO.
  • Suitable for receptor binding, signaling, and antiviral assays.