Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC It1t dihydrochloride | 1092776-63-0 | MFCD22683853 | ≥98.0% | 479.57 g/mol | C21H36Cl2N4S2 | 10 MG
SDP

Catalog No. 5000249093
Encompass_Preferred

IT1t dihydrochloride is a small-molecule antagonist of the chemokine receptor CXCR4, used in research to study CXCL12/CXCR4 signaling and ligand binding. It inhibits the CXCL12-CXCR4 interaction with a reported IC50 of about 2.1 nM and is provided as the dihydrochloride salt for improved stability.

  • Potent CXCR4 antagonist with IC50 ≈2.1 nM.
  • Suitable for receptor binding and functional assays.
  • Supplied as the dihydrochloride salt to improve stability.
  • Soluble in water (≈50 mg/mL) and in DMSO (≥30 mg/mL).
  • Molecular formula C21H36Cl2N4S2; molecular weight 479.57 g/mol.
  • High reported purity (≥95%-≥98% depending on supplier).
  • Typical research pack size 10 MG.

Catalog No. 50-002-49093 Supplier Medchemexpress LLC Supplier No. HY101458A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

IT1t dihydrochloride is a small-molecule antagonist of the chemokine receptor CXCR4, used in research to study CXCL12/CXCR4 signaling and ligand binding. It inhibits the CXCL12-CXCR4 interaction with a reported IC50 of about 2.1 nM and is provided as the dihydrochloride salt for improved stability.

  • Potent CXCR4 antagonist with IC50 ≈2.1 nM.
  • Suitable for receptor binding and functional assays.
  • Supplied as the dihydrochloride salt to improve stability.
  • Soluble in water (≈50 mg/mL) and in DMSO (≥30 mg/mL).
  • Molecular formula C21H36Cl2N4S2; molecular weight 479.57 g/mol.
  • High reported purity (≥95%-≥98% depending on supplier).
  • Typical research pack size 10 MG.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.