Learn More
Medchemexpress LLC IT1t dihydrochloride | 1092776-63-0 | MFCD22683853 | 99.8% | 479.57 | C21H36Cl2N4S2 | 50 MG
SDP
Catalog No. 5000247950
Encompass_Preferred
IT1t dihydrochloride is a potent small-molecule antagonist of the CXCR4 chemokine receptor. It inhibits CXCL12/CXCR4 interaction with high potency (IC50 ≈ 2.1 nM), shows activity in HIV-1 (X4) cellular assays, and has been used in preclinical metastasis models. Supplied as the dihydrochloride salt for research use.
  • Potent CXCR4 antagonist with low-nanomolar potency.
  • Active in HIV-1 (X4) cellular assays.
  • Suitable for CXCR4 signaling and metastasis studies.
  • Provided as a dihydrochloride salt for stability.
  • High reported purity (≈99.8%).
  • Available in small research pack sizes (mg scale).
Catalog No. 50-002-47950 Supplier Medchemexpress LLC Supplier No. HY101458A50MG
IT1t dihydrochloride is a potent small-molecule antagonist of the CXCR4 chemokine receptor. It inhibits CXCL12/CXCR4 interaction with high potency (IC50 ≈ 2.1 nM), shows activity in HIV-1 (X4) cellular assays, and has been used in preclinical metastasis models. Supplied as the dihydrochloride salt for research use.
  • Potent CXCR4 antagonist with low-nanomolar potency.
  • Active in HIV-1 (X4) cellular assays.
  • Suitable for CXCR4 signaling and metastasis studies.
  • Provided as a dihydrochloride salt for stability.
  • High reported purity (≈99.8%).
  • Available in small research pack sizes (mg scale).