Learn More
Enzo Life Sciences ITAC monoclonal antibody (22F1) (biotin conjugate) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZABS2750100
Encompass_Preferred
Clone: 22F1. Isotype: IgG1. Immunogen: Full length recombinant ITAC produced in E. coli. Species reactivity: Human. Applications: ELISA, WB. Purity: Protein G affinity purified. Formulation: Liquid. In PBS containing 4% BSA and a proprietary preservative.
Catalog No. 50-201-2819