Learn More
Medchemexpress LLC Itacitinib | 1334298-90-6 | 100.0% | 553.51 | 500 MG
SDP
Catalog No. 5000391328
Encompass_Preferred
Itacitinib (INCB039110) is an orally active and selective inhibitor of JAK1 with an IC50 of 2 nM for human JAK1. Itacitinib shows >20-fold selectivity for JAK1 over JAK2 and >100-fold over JAK3 and TYK2; Itacitinib is used in the research of myelofibrosis.
  • Orally active and selective inhibitor of JAK1
  • IC50 of 2 nM for human JAK1
  • >20-fold selectivity for JAK1 over JAK2
  • >100-fold selectivity for JAK1 over JAK3 and TYK2
  • Used in the research of myelofibrosis
Catalog No. 50-003-91328 Supplier Medchemexpress LLC Supplier No. HY16997500MG
Itacitinib (INCB039110) is an orally active and selective inhibitor of JAK1 with an IC50 of 2 nM for human JAK1. Itacitinib shows >20-fold selectivity for JAK1 over JAK2 and >100-fold over JAK3 and TYK2; Itacitinib is used in the research of myelofibrosis.
  • Orally active and selective inhibitor of JAK1
  • IC50 of 2 nM for human JAK1
  • >20-fold selectivity for JAK1 over JAK2
  • >100-fold selectivity for JAK1 over JAK3 and TYK2
  • Used in the research of myelofibrosis