Learn More
Medchemexpress LLC Itacitinib | 1334298-90-6 | 99.97% | 553.51 | 25 MG
SDP
Catalog No. 5000391972
Encompass_Preferred
Itacitinib is an orally active and selective inhibitor of JAK1 with an IC50 of 2 nM for human JAK1. It shows >20-fold selectivity for JAK1 over JAK2 and >100-fold over JAK3 and TYK2. Itacitinib is used in the research of myelofibrosis.
  • Orally active
  • Selective inhibitor of JAK1
  • High selectivity over JAK2, JAK3, and TYK2
  • Used in the research of myelofibrosis
Catalog No. 50-003-91972 Supplier Medchemexpress LLC Supplier No. 25MGHY16997
Itacitinib is an orally active and selective inhibitor of JAK1 with an IC50 of 2 nM for human JAK1. It shows >20-fold selectivity for JAK1 over JAK2 and >100-fold over JAK3 and TYK2. Itacitinib is used in the research of myelofibrosis.
  • Orally active
  • Selective inhibitor of JAK1
  • High selectivity over JAK2, JAK3, and TYK2
  • Used in the research of myelofibrosis