Learn More
TARGETMOL CHEMICALS INC Itacitinib 5MG
SDP
Catalog No. 502880440
Encompass_Preferred
Also available in 1 mL, 1 mg, 2 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Itacitinib (INCB039110) is an orally bioavailable Janus-associated kinase 1 (JAK1) inhibitor with potential antineoplastic activity. Purity 97.57%
Catalog No. 50-288-0440 Supplier TARGETMOL CHEMICALS INC Supplier No. T39985MG
Also available in 1 mL, 1 mg, 2 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Itacitinib (INCB039110) is an orally bioavailable Janus-associated kinase 1 (JAK1) inhibitor with potential antineoplastic activity. Purity 97.57%