Learn More
Medchemexpress LLC Itacitinib adipate | 1334302-63-4 | 99.2% | 699.66 | 1 ML
SDP
Catalog No. 5000393915
Encompass_Preferred
Itacitinib adipate is an orally bioavailable and selective JAK1 inhibitor which has been tested for efficacy and safety in a phase II trial in myelofibrosis.
  • Orally bioavailable and selective JAK1 inhibitor.
  • Tested for efficacy and safety in a phase II trial in myelofibrosis.
  • For research use only.
Catalog No. 50-003-93915 Supplier Medchemexpress LLC Supplier No. HY16997A10MM/1ML
Itacitinib adipate is an orally bioavailable and selective JAK1 inhibitor which has been tested for efficacy and safety in a phase II trial in myelofibrosis.
  • Orally bioavailable and selective JAK1 inhibitor.
  • Tested for efficacy and safety in a phase II trial in myelofibrosis.
  • For research use only.