Learn More
Medchemexpress LLC Itacitinib adipate | 1334302-63-4 | 99.2% | 699.66 | 5 MG
SDP
Catalog No. 5000393916
Encompass_Preferred
Itacitinib adipate is an orally bioavailable and selective JAK1 inhibitor that has been tested for efficacy and safety in a phase II trial in myelofibrosis. It is suitable for research use only.
  • Orally bioavailable
  • Selective JAK1 inhibitor
  • Tested in phase II trial for myelofibrosis
  • Suitable for research use
Catalog No. 50-003-93916 Supplier Medchemexpress LLC Supplier No. HY16997A5MG
Itacitinib adipate is an orally bioavailable and selective JAK1 inhibitor that has been tested for efficacy and safety in a phase II trial in myelofibrosis. It is suitable for research use only.
  • Orally bioavailable
  • Selective JAK1 inhibitor
  • Tested in phase II trial for myelofibrosis
  • Suitable for research use