Learn More
Medchemexpress LLC Itacitinib adipate | 1334302-63-4 | 99.2% | 699.66 | 50 MG
SDP
Catalog No. 5000393579
Encompass_Preferred
Itacitinib adipate is an orally bioavailable and selective JAK1 inhibitor which has been tested for efficacy and safety in a phase II trial in myelofibrosis.
  • Orally bioavailable and selective JAK1 inhibitor.
  • Evaluated in phase II clinical trials for myelofibrosis.
  • Available in various quantities and solution formats.
  • High purity.
  • Involved in numerous clinical trials for various conditions including Acute Lymphoblastic Leukemia, Systemic Sclerosis, Graft-versus-host Disease, and various cancers.
Catalog No. 50-003-93579 Supplier Medchemexpress LLC Supplier No. HY16997A50MG
Itacitinib adipate is an orally bioavailable and selective JAK1 inhibitor which has been tested for efficacy and safety in a phase II trial in myelofibrosis.
  • Orally bioavailable and selective JAK1 inhibitor.
  • Evaluated in phase II clinical trials for myelofibrosis.
  • Available in various quantities and solution formats.
  • High purity.
  • Involved in numerous clinical trials for various conditions including Acute Lymphoblastic Leukemia, Systemic Sclerosis, Graft-versus-host Disease, and various cancers.