Learn More
Selleck Chemical LLC Itacitinib (INCB39110) 2mg 1334298-90-6
SDP
Supplier:  Selleck Chemical LLC S78122MG
Encompass_Preferred
Itacitinib(INCB39110) is an orally bioavailable inhibitor of Janus-associated kinase 1 (JAK1) with potential antineoplastic activity. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8171