Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Itacnosertib (hydrochloride) | 2409543-84-4 | 99.4% | 505.01 | 50 MG
SDP

Catalog No. 5000344517
Encompass_Preferred

Itacnosertib hydrochloride (TP-0184 hydrochloride) is an inhibitor for FLT3, ACVR1, and JAK2. It exhibits anti-leukemic activity and is intended for research use only. This compound has shown promising results in pre-clinical studies, demonstrating its potential in various biological applications.

  • Inhibits FLT3, ACVR1 (ALK2), and JAK2
  • Exhibits anti-leukemic activity
  • Arrests cell cycle of MOLM-13, MOLM-14, and MV4-11 at G1/G0 phase
  • Inhibits proliferation of acute myeloid leukemia (AML) cells
  • Inhibits activation of FLT3 and ACVR1 downstream signaling pathways
  • Shows antitumor activity in mouse xenograft models

Catalog No. 50-003-44517 Supplier Medchemexpress LLC Supplier No. HY109179A10MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Itacnosertib hydrochloride (TP-0184 hydrochloride) is an inhibitor for FLT3, ACVR1, and JAK2. It exhibits anti-leukemic activity and is intended for research use only. This compound has shown promising results in pre-clinical studies, demonstrating its potential in various biological applications.

  • Inhibits FLT3, ACVR1 (ALK2), and JAK2
  • Exhibits anti-leukemic activity
  • Arrests cell cycle of MOLM-13, MOLM-14, and MV4-11 at G1/G0 phase
  • Inhibits proliferation of acute myeloid leukemia (AML) cells
  • Inhibits activation of FLT3 and ACVR1 downstream signaling pathways
  • Shows antitumor activity in mouse xenograft models

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.