Learn More
Medchemexpress LLC Itacnosertib hydrochloride | 2409543-84-4 | 99.4% | 505.01 | C26H29ClN8O | 5 MG
SDP
Catalog No. 5000263634
Encompass_Preferred
Itacnosertib hydrochloride is the hydrochloride salt of TP-0184, a small-molecule investigational inhibitor that potently targets ACVR1/ALK2 (reported IC50 ≈ 8 nM), shows activity against FLT3, and has markedly weaker activity versus JAK2 (reported IC50 ≈ 8,540 nM). It has demonstrated anti-leukemic activity in preclinical studies and is supplied for research use as a high-purity hydrochloride salt.
  • Potent ACVR1/ALK2 inhibition (IC50 ≈ 8 nM).
  • Activity against FLT3 with reported anti-leukemic effects.
  • Markedly weaker JAK2 activity (IC50 ≈ 8,540 nM), indicating selectivity.
  • Supplied as the hydrochloride salt for improved solubility.
  • High reported purity (~99.4%) backed by certificate of analysis.
  • Provided in small research pack sizes suitable for in vitro studies.
Catalog No. 50-002-63634 Supplier Medchemexpress LLC Supplier No. HY109179A5MG
Itacnosertib hydrochloride is the hydrochloride salt of TP-0184, a small-molecule investigational inhibitor that potently targets ACVR1/ALK2 (reported IC50 ≈ 8 nM), shows activity against FLT3, and has markedly weaker activity versus JAK2 (reported IC50 ≈ 8,540 nM). It has demonstrated anti-leukemic activity in preclinical studies and is supplied for research use as a high-purity hydrochloride salt.
  • Potent ACVR1/ALK2 inhibition (IC50 ≈ 8 nM).
  • Activity against FLT3 with reported anti-leukemic effects.
  • Markedly weaker JAK2 activity (IC50 ≈ 8,540 nM), indicating selectivity.
  • Supplied as the hydrochloride salt for improved solubility.
  • High reported purity (~99.4%) backed by certificate of analysis.
  • Provided in small research pack sizes suitable for in vitro studies.