Learn More
Medchemexpress LLC Itacnosertib hydrochloride (TP-0184 hydrochloride) | 2409543-84-4 | 99.4% | 505.01 | 1 ML
SDP
Catalog No. 5000344518
Encompass_Preferred
Itacnosertib hydrochloride (TP-0184 hydrochloride) is an inhibitor for FLT3, ACVR1 (ALK2, IC50=8 nM), and JAK2 (IC50=8540 nM) that exhibits anti-leukemic activity.
  • Inhibits FLT3, ACVR1 (ALK2), and JAK2.
  • Exhibits anti-leukemic activity.
  • Arrests the cell cycle of MOLM-13, MOLM-14, and MV4-11 at G1/G0 phase.
  • Inhibits the proliferation of acute myeloid leukemia (AML) cells.
  • Inhibits the activation of FLT3 and ACVR1 downstream signaling pathways.
  • Inhibits serine biosynthesis and amino acid transport pathways.
  • Exhibits antitumor activity against MOLM-13 in mouse models.
Catalog No. 50-003-44518 Supplier Medchemexpress LLC Supplier No. HY109179A10MG
Itacnosertib hydrochloride (TP-0184 hydrochloride) is an inhibitor for FLT3, ACVR1 (ALK2, IC50=8 nM), and JAK2 (IC50=8540 nM) that exhibits anti-leukemic activity.
  • Inhibits FLT3, ACVR1 (ALK2), and JAK2.
  • Exhibits anti-leukemic activity.
  • Arrests the cell cycle of MOLM-13, MOLM-14, and MV4-11 at G1/G0 phase.
  • Inhibits the proliferation of acute myeloid leukemia (AML) cells.
  • Inhibits the activation of FLT3 and ACVR1 downstream signaling pathways.
  • Inhibits serine biosynthesis and amino acid transport pathways.
  • Exhibits antitumor activity against MOLM-13 in mouse models.