Learn More
Medchemexpress LLC Itaconate-alkyne | 2454181-83-8 | 98.0% | 238.28 g/mol | C13H18O4 | 5 MG
SDP
Catalog No. 5000263095
Encompass_Preferred
Itaconate-alkyne is a bioorthogonal alkyne probe used for quantitative, site-specific chemoproteomic profiling of itaconation in living cells. As a functional analogue of itaconate, it enables click-chemistry labeling of itaconylated proteins to support target identification and downstream biochemical analysis.
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAC) labeling
  • Designed for quantitative, site-specific chemoproteomic workflows
  • Functional analogue of itaconate for biologically relevant target capture
  • White to off-white solid suitable for formulation in common organic solvents
  • High purity for consistent experimental results
  • Stable when stored at recommended low temperatures
Catalog No. 50-002-63095 Supplier Medchemexpress LLC Supplier No. HY1338705MG
Itaconate-alkyne is a bioorthogonal alkyne probe used for quantitative, site-specific chemoproteomic profiling of itaconation in living cells. As a functional analogue of itaconate, it enables click-chemistry labeling of itaconylated proteins to support target identification and downstream biochemical analysis.
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAC) labeling
  • Designed for quantitative, site-specific chemoproteomic workflows
  • Functional analogue of itaconate for biologically relevant target capture
  • White to off-white solid suitable for formulation in common organic solvents
  • High purity for consistent experimental results
  • Stable when stored at recommended low temperatures