Learn More
Medchemexpress LLC Itaconate-alkyne | 2454181-83-8 | 98.0% | C13H18O4 | 10MG
SDP
Catalog No. 5000259476
Encompass_Preferred
Itaconate-alkyne is an alkyne-tagged bioorthogonal probe designed for quantitative and site-specific chemoproteomic profiling of itaconation in living cells. As a functional analogue of itaconate, it enables labeling of bona fide itaconate targets and is compatible with click-chemistry workflows.
  • Contains alkyne group for copper-catalyzed azide-alkyne cycloaddition.
  • Enables quantitative and site-specific chemoproteomic profiling.
  • Functions as a structural analogue of itaconate with comparable anti-inflammatory effects.
  • High purity solid form for reliable experimental results.
  • Available in multiple small-scale packaging sizes suitable for laboratory use.
  • Compatible with standard click-chemistry reagents and protocols.
Catalog No. 50-002-59476 Supplier Medchemexpress LLC Supplier No. HY13387010MG
Itaconate-alkyne is an alkyne-tagged bioorthogonal probe designed for quantitative and site-specific chemoproteomic profiling of itaconation in living cells. As a functional analogue of itaconate, it enables labeling of bona fide itaconate targets and is compatible with click-chemistry workflows.
  • Contains alkyne group for copper-catalyzed azide-alkyne cycloaddition.
  • Enables quantitative and site-specific chemoproteomic profiling.
  • Functions as a structural analogue of itaconate with comparable anti-inflammatory effects.
  • High purity solid form for reliable experimental results.
  • Available in multiple small-scale packaging sizes suitable for laboratory use.
  • Compatible with standard click-chemistry reagents and protocols.