Learn More
TARGETMOL CHEMICALS INC ITACONATE-ALKYNE 5MG
SDP
Catalog No. 502579937
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Itaconate-alkyne (ITalk) is a bioorthogonal probe for quantitative and site-specific chemoproteomic profiling of itaconate in inflammatory macrophages and enables biochemical evaluation and proteomic analysis of its direct targets. purity: 99%
Catalog No. 50-257-9937 Supplier TARGETMOL CHEMICALS INC Supplier No. T413025MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Itaconate-alkyne (ITalk) is a bioorthogonal probe for quantitative and site-specific chemoproteomic profiling of itaconate in inflammatory macrophages and enables biochemical evaluation and proteomic analysis of its direct targets. purity: 99%