Learn More
Medchemexpress LLC HY-Y0520 100mg Medchemexpress, Itaconic acid CAS:97-65-4 Purity:>98%
Supplier: Medchemexpress LLC HYY0520100MG
Medchemexpress, HY-Y0520 100mg Itaconic acid CAS:97-65-4 Itaconic acid, a precursor of polymers, chemicals, and fuels, can be synthesized by many fungi. Itaconic acid also is a macrophage-specific metabolite. Itaconic acid mediates crosstalk between macrophage metabolism and peritoneal tumors. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.