Learn More
Medchemexpress LLC Itaconic acid-13C5 | 2095777-38-9 | 97.0% | 135.06 | 13C5H6O4 | 50ug
SDP
Catalog No. 5000268886
Encompass_Preferred
Itaconic acid-13C5 is the 13C labeled Itaconic acid Itaconic acid a precursor of polymers chemicals and fuels can be synthesized by many fungi Itaconic acid also is a macrophage-specific metabolite Itaconic acid mediates crosstalk between macrophage metabolism and peritoneal tumors[1][2]
Catalog No. 50-002-68886 Supplier Medchemexpress LLC Supplier No. HYY0520S50UG
Itaconic acid-13C5 is the 13C labeled Itaconic acid Itaconic acid a precursor of polymers chemicals and fuels can be synthesized by many fungi Itaconic acid also is a macrophage-specific metabolite Itaconic acid mediates crosstalk between macrophage metabolism and peritoneal tumors[1][2]