Learn More
Selleck Chemical LLC Itaconic acid S3095-1g
SDP
Catalog No. 503111312
Encompass_Preferred
Itaconic acid a macrophage-specific (M -specific) metabolite mediates crosstalk between macrophage metabolism and peritoneal tumors Itaconic acid is also a precursor of polymers chemicals and fuels that can be synthesized by many fungi
Catalog No. 50-311-1312 Supplier Selleck Chemical LLC Supplier No. S30951G
Itaconic acid a macrophage-specific (M -specific) metabolite mediates crosstalk between macrophage metabolism and peritoneal tumors Itaconic acid is also a precursor of polymers chemicals and fuels that can be synthesized by many fungi