Learn More
TARGETMOL CHEMICALS INC Itaconic acid 500MG
SDP
Catalog No. 502878676
Encompass_Preferred
Also available in 1 g, 1 mL, 100 mg and bulk. Please contact Fisher for quotes. Itaconic acid is a precursor for the synthesis of polymers, chemicals, and fuels, and has anti-inflammatory activity.Itaconic acid can be synthesized by fungi. It is also a macrophage-specific metabolite. It mediates crosstalk between macrophage metabolism and peritoneal tumors. Purity 100%
Catalog No. 50-287-8676 Supplier TARGETMOL CHEMICALS INC Supplier No. T4837500MG
Also available in 1 g, 1 mL, 100 mg and bulk. Please contact Fisher for quotes. Itaconic acid is a precursor for the synthesis of polymers, chemicals, and fuels, and has anti-inflammatory activity.Itaconic acid can be synthesized by fungi. It is also a macrophage-specific metabolite. It mediates crosstalk between macrophage metabolism and peritoneal tumors. Purity 100%