Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Itaconic acid, ≥96.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111015998 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5998 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5998 Supplier MilliporeSigma Supelco Supplier No. 93598100MG
Add to Cart
Add to Cart

Itaconic acid, an unsaturated dicarbonic organic acid, is a metabolic product of Aspergillus itaconicus.

TRUSTED_SUSTAINABILITY

Specifications

Ignition Point 1472°F
CAS 97-65-4
Density 1.573 g/mL (at 25°C)
Physical Form Neat
Linear Formula HO2 CCH2 C(=CH2)CO2 H
MDL Number MFCD00004260
Melting Point 165°C to 168°C
Synonym Methylenesuccinic acid
Type Analytical Standard
Quantity 100 mg
Formula Weight 130.10
Percent Purity ≥96.0% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.