Learn More
Apexbio Technology LLC Itaconic acid 97-65-4 500mg
SDP
Catalog No. 5000091188
Encompass_Preferred
Itaconic acid (CAS 97-65-4) is a dicarboxylic acid commonly investigated for its role as a monomer in biomedical research Its incorporation in the polymerization processes such as with polyacrylonitrile (PAN) facilitates the thermo-oxidative stabilization of resulting polymers by promoting cross-linking and enhancing structural integrity under oxidative conditions This property is significant in the development of advanced biomaterials including tissue engineering scaffolds and drug delivery systems Itaconic acid s chemical reactivity and compatibility with various polymer matrices make it valuable for studies focusing on material modification and functionalization
Catalog No. 50-000-91188 Supplier Apexbio Technology LLC Supplier No. B7891500
Itaconic acid (CAS 97-65-4) is a dicarboxylic acid commonly investigated for its role as a monomer in biomedical research Its incorporation in the polymerization processes such as with polyacrylonitrile (PAN) facilitates the thermo-oxidative stabilization of resulting polymers by promoting cross-linking and enhancing structural integrity under oxidative conditions This property is significant in the development of advanced biomaterials including tissue engineering scaffolds and drug delivery systems Itaconic acid s chemical reactivity and compatibility with various polymer matrices make it valuable for studies focusing on material modification and functionalization