Learn More
Medchemexpress LLC Itaconic acid | 97-65-4 | MFCD00004260 | 99.9% | 130.10 | C5H6O4 | 1mL
SDP
Catalog No. 5000269070
Encompass_Preferred
Itaconic acid a precursor of polymers chemicals and fuels can be synthesized by many fungi Itaconic acid also is a macrophage-specific metabolite Itaconic acid mediates crosstalk between macrophage metabolism and peritoneal tumors Itaconic acid has anti-inflammatory anti-microbial and immunomodulatory effect[1][2][4][5]
Catalog No. 50-002-69070 Supplier Medchemexpress LLC Supplier No. HYY052010MM/1ML
Itaconic acid a precursor of polymers chemicals and fuels can be synthesized by many fungi Itaconic acid also is a macrophage-specific metabolite Itaconic acid mediates crosstalk between macrophage metabolism and peritoneal tumors Itaconic acid has anti-inflammatory anti-microbial and immunomodulatory effect[1][2][4][5]