Learn More
AdipoGen Itaconic acid Itaconate
SDP
Catalog No. 502562534
Encompass_Preferred
Chemical. CAS 97-65-4. Formula C5H6O4. MW 130.1. Mammalian antibacterial metabolite. Inhibitor of isocitrate lyase, the key enzyme of the glyoxylate cycle, essential for bacterial growth. Antimicrobial. Antibacterial. Inhibits the growth of bacteria expressing isocitrate lyase. Phosphofructokinase PFKII inhibitor. Suppresses glycolysis by decreasing the level of fructose 2,6-bisphosphate, resulting in decreased visceral fat. Might be involved in the regulation of metabolism. One of the most highly induced metabolites in activated macrophages. Exerts anti-inflammatory effects in vitro and in vivo. Regulates succinate levels by inhibiting succinate dehydrogenase SDH. Regulates mitochondrial respiration and cytokine production. Might induce inflammasome activation and consequently IL-1beta secretion by generating ROS.
Catalog No. 50-256-2534 Supplier AdipoGen Supplier No. AGCN20426G005
Chemical. CAS 97-65-4. Formula C5H6O4. MW 130.1. Mammalian antibacterial metabolite. Inhibitor of isocitrate lyase, the key enzyme of the glyoxylate cycle, essential for bacterial growth. Antimicrobial. Antibacterial. Inhibits the growth of bacteria expressing isocitrate lyase. Phosphofructokinase PFKII inhibitor. Suppresses glycolysis by decreasing the level of fructose 2,6-bisphosphate, resulting in decreased visceral fat. Might be involved in the regulation of metabolism. One of the most highly induced metabolites in activated macrophages. Exerts anti-inflammatory effects in vitro and in vivo. Regulates succinate levels by inhibiting succinate dehydrogenase SDH. Regulates mitochondrial respiration and cytokine production. Might induce inflammasome activation and consequently IL-1beta secretion by generating ROS.