Learn More
BIOHIPPO Itaconyl-Cysteine Rabbit mAb
SDP
Catalog No. 5000975559
Encompass
U-Blot Pan-Itaconyl-Cysteine Rabbit mAb is a Rabbit-derived monoclonal antibody that specifically recognizes Itaconyl-Cysteine a cysteine itaconylation modification at Cys(C) Itaconyl-Cysteine (C(itacon)) is a novel metabolism-dependent post-translational modification occurring on cysteine residues of proteins This modification is mediated by itaconate a metabolite abundantly produced under immune activation and inflammatory conditions via covalent reaction with the thiol group of cysteine Itaconylation is widely involved in key biological processes including inflammatory signaling oxidative stress response innate immunity and metabolic reprogramming It plays critical regulatory roles in the development of inflammatory diseases infectious immunity and metabolism-related disorders serving as an important molecular marker and research target linking immunometabolism and protein function regulation Supplied as 100 uL
Catalog No. 50-009-75559 Supplier BIOHIPPO Supplier No. BHA21821318
U-Blot Pan-Itaconyl-Cysteine Rabbit mAb is a Rabbit-derived monoclonal antibody that specifically recognizes Itaconyl-Cysteine a cysteine itaconylation modification at Cys(C) Itaconyl-Cysteine (C(itacon)) is a novel metabolism-dependent post-translational modification occurring on cysteine residues of proteins This modification is mediated by itaconate a metabolite abundantly produced under immune activation and inflammatory conditions via covalent reaction with the thiol group of cysteine Itaconylation is widely involved in key biological processes including inflammatory signaling oxidative stress response innate immunity and metabolic reprogramming It plays critical regulatory roles in the development of inflammatory diseases infectious immunity and metabolism-related disorders serving as an important molecular marker and research target linking immunometabolism and protein function regulation Supplied as 100 uL