Learn More
Medchemexpress LLC HY-14399 5mg Medchemexpress, Itanapraced CAS:749269-83-8 Purity:>98%
SDP
Catalog No. 502081597
Encompass_Preferred
Medchemexpress, HY-14399 5mg Itanapraced CAS:749269-83-8 Itanapraced (CHF5074) is a novel γ-secretase modulator, reduces Aβ42 and Aβ40 secretion, with an IC50 of 3.6 and 18.4 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1597 Supplier Medchemexpress LLC Supplier No. HY143995MG
Medchemexpress, HY-14399 5mg Itanapraced CAS:749269-83-8 Itanapraced (CHF5074) is a novel γ-secretase modulator, reduces Aβ42 and Aβ40 secretion, with an IC50 of 3.6 and 18.4 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.