Learn More
Bio Rad Laboratories ITAQ UNIVER SYBR GREEN 2ML 2X1
Catalog No. NC1720486
Encompass
Supermix, 200 x 20 µl rxns, 2 ml (2 x 1 ml)
Catalog No. NC1720486 Supplier Bio Rad Laboratories Supplier No. 1725120
Supermix, 200 x 20 µl rxns, 2 ml (2 x 1 ml)