Learn More
Bio Rad Laboratories iTaq Universal SYBR Green Supermix, 500 x 20 uL rxns, 5 mL (5 x 1 mL)
Catalog No. NC1085841
Encompass
500 x 20 uL reactions, 5 mL (5 x 1 mL), 2x qPCR mix, contains dNTPs, iTaq DNA Polymerase, MgCl2, SYBR Green I, enhancers, stabilizers, fluorescein, and ROX Normalization Dyes.
Catalog No. NC1085841 Supplier Bio Rad Laboratories Supplier No. 1725121
500 x 20 uL reactions, 5 mL (5 x 1 mL), 2x qPCR mix, contains dNTPs, iTaq DNA Polymerase, MgCl2, SYBR Green I, enhancers, stabilizers, fluorescein, and ROX Normalization Dyes.