Learn More
TARGETMOL CHEMICALS INC ITASETRON 5MG
SDP
Catalog No. 502580824
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Itasetron (U 98079A) is a 5-hydroxytryptamine 3-receptor antagonist used to prevent age-related memory deficits in rats.Itasetron's antiemetic activity in animal models of cancer chemotherapy and radiation therapy may be useful in the study of amnesia. purity: 99%
Catalog No. 50-258-0824 Supplier TARGETMOL CHEMICALS INC Supplier No. T241845MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Itasetron (U 98079A) is a 5-hydroxytryptamine 3-receptor antagonist used to prevent age-related memory deficits in rats.Itasetron's antiemetic activity in animal models of cancer chemotherapy and radiation therapy may be useful in the study of amnesia. purity: 99%