Learn More
Optimize Technologies ITB™ PTFE Plunger Seal, Waters 225 µL Head (#3) [510, 590, 600/600E, 610, 650, 6K/6KA]
SDP
Catalog No. 50114188
Encompass_Preferred
Optimize Piston Seals are the longest lasting seals on the market. Machined from either of two proprietary polymer blends, UHMW-PE or ITB, Optimize Seals provide the ultimate in high pressure sealing capacity. For mostly aqueous buffered mobile phases, PTFE-based Improved PTFE Blend (ITB) seals are a superior choice for applications with elevated temperatures, high organic content, or when the broadest possible solvent compatibility is required. ITB™ seals tend to be softer than those made from UHMW-PE, making them more forgiving at high pressures where piston side-loading or misalignment may occur. This softer nature also leads to more shedding, but the ITB™ proprietary blend is designed to shed particles that are large enough to be caught at the surface of a standard 2µm frit, not embed within it. This item is equivalent to OEM part number WAT026644.
Catalog No. 50-114-188 Supplier Optimize Technologies Supplier No. 393601214
Optimize Piston Seals are the longest lasting seals on the market. Machined from either of two proprietary polymer blends, UHMW-PE or ITB, Optimize Seals provide the ultimate in high pressure sealing capacity. For mostly aqueous buffered mobile phases, PTFE-based Improved PTFE Blend (ITB) seals are a superior choice for applications with elevated temperatures, high organic content, or when the broadest possible solvent compatibility is required. ITB™ seals tend to be softer than those made from UHMW-PE, making them more forgiving at high pressures where piston side-loading or misalignment may occur. This softer nature also leads to more shedding, but the ITB™ proprietary blend is designed to shed particles that are large enough to be caught at the surface of a standard 2µm frit, not embed within it. This item is equivalent to OEM part number WAT026644.