Learn More
Aviva Systems Biology Corp ITCH Antibody - N-terminal region (ARP43347_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43347_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ITCH. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ITCH. Alternate names: AIF4, AIP4, NAPP1, dJ468O1.1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Rabbit, Rat
Catalog No. 50-014-58504