Learn More
Selleck Chemical LLC ITD-1-5mg
SDP
Supplier:  Selleck Chemical LLC S67135MG
Encompass_Preferred
ITD-1 is a potent TGF-β inhibitor. It does not block the kinase activity of either type I (TGFBR1) or type II (TGFBR2) TGFβ receptors but potently blocks phosphorylation of the effector SMAD2/3 proteins induced by TGFβ2, and only minimally in response to Activin A.
Catalog No. 50-238-9898