Learn More
Pyramex™ Itek™ Safety Eyewear
Catalog No. 19155598 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Lens Coating:
Anti-fog
Uncoated
Lens Tint:
Amber
Blue
Blue, Mirror
Clear
Gray
Indoor/Outdoor, Mirror
Silver, Mirror
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Coating Lens Tint
19-155-598 Uncoated Indoor/Outdoor, Mirror
19-155-590 Uncoated Clear
19-155-591 Anti-fog Clear
19-155-592 - Gray
19-155-593 Anti-fog Gray
19-155-594 - Amber
19-155-595 Uncoated Blue
19-155-596 Uncoated Silver, Mirror
19-155-597 - Blue, Mirror
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Catalog No. 19-155-598 Supplier Pyramex™ Supplier No. S5880S
Add to Cart
Add to Cart
This item is not returnable. View return policy
Lightweight, frameless protection is ideal for all-day wear
  • Soft integrated nose piece
  • Soft dual injected rubber temple tips provide non-slip comfort
  • 9.75 base curve lens provides excellent side protection
  • Scratch resistant polycarbonate lens provides 99% UVA/B/C protection
  • Passes MIL-PRF 32432 High Velocity Impact Standardstrap
  • Specifications
    Lens Coating Uncoated
    Certifications/Compliance ANSI Z87.1, CSA Z94.3-07
    Certifications/Compliance (Europe) EN-166
    Lens Tint Indoor/Outdoor, Mirror
    Weight (Metric) 23.5 g
    Lens Material Polycarbonate