Learn More
Electron Microscopy Sciences Bench Top Small Item Organizer, 4 Levels, 8 Bins
SDP
Catalog No. 5030581
Encompass_Preferred
This organizer has adjustable dividers that allow you to change compartment size to fit the items including compact sizes for storing small items. It is made from soild PVC, chemically resistant, and very light weight. Comes without door. Measures: 12"(L)x11"(D)x20"(H).
Catalog No. 50-305-81 Supplier Electron Microscopy Sciences Supplier No. 6438408
This organizer has adjustable dividers that allow you to change compartment size to fit the items including compact sizes for storing small items. It is made from soild PVC, chemically resistant, and very light weight. Comes without door. Measures: 12"(L)x11"(D)x20"(H).