Learn More
Apexbio Technology LLC ITF2357 (Givinostat),5mg CAS# 732302-99-7
SDP
Supplier:  Apexbio Technology LLC A40935
Encompass_Preferred
ITF2357, also known as givinostat, is a potent inhibitor of both class I and class II histone deacetylase (HDAC) as well as a potent inhibitor of hematopoietic colony formation by JAKEV617F-bearing progenitor cells from chronic myeloproliferative neoplasms in vitro. Previous studies has shown that ITF2357 induces apoptosis of multiple myeloma (MM) and acute myelogenous leukemia (AML) cells following induction of p21 and down-modulation of Bcl-2 and Mcl-1 proteins and inhibits the production of pro-inflammatory cytokines (such as IL-1, IL-6, tumor necrosis factor (TNF)-α and interferon (IFN)-γ) by peripheral blood mononuclear cells as well as the production of IL-6 and vascular endothelium growth factor (VEGF) by mesenchymal stromal cells. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-224-0017