Learn More
Aviva Systems Biology Corp ITGA2 Antibody - C-terminal region (ARP64290_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64290_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ITGA2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C-terminal region of Human ITGA2. Alternate names: BR, CD49B, GPIa, VLA-2, VLAA2, BDPLT9ApplicationIF, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-288-223