Learn More
Aviva Systems Biology Corp ITGA5 Antibody - C-terminal region (ARP64613_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64613_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ITGA5. It was validated on Western Blot. . Alternate names: CD49e, FNRA, VLA5AApplicationWB. Species reactivity: Human, Mouse
Catalog No. 50-288-315