Learn More
Biomatik Corporation Itga6 Polyclonal Antibody
Supplier:  Biomatik Corporation CAC11194
Encompass_Preferred
Itga6 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to ITGA6. The Itga6 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Integrin Alpha-6 Protein (625-866Aa). Applications: ELISA, WB, IF. Target Synonyms: ITGA6. Accession Number: P23229. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6114