Learn More
Aviva Systems Biology Corp ITGB3BP Antibody - N-terminal region (ARP54999_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54999_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ITGB3BP. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ITGB3BP. Alternate names: CENP-R, CENPR, HSU37139, NRIF3, TAP20ApplicationWB. Species reactivity: Human
Catalog No. 50-014-61393